Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "vacuum-diaphragm-pumps"

https://nz-bot.vwr.com/stibo/smallweb/std.lang.all/99/72/18989972.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalogue Number: (BUCH071000)
Supplier:  BÜCHI
Local Article Number:: BUCH071000
Description:   The V-700 vacuum pump is a chemically resistant PTFE diaphragm pump designed for use with all gases and vapours typically associated in the lab. With ...
UOM:  1 * 1 items
Catalogue Number: (BUCH11V300100)
Supplier:  BÜCHI
Local Article Number:: BUCH11V300100
Description:   The chemically resistant PTFE diaphragm vacuum pump impresses with its very silent and economical operation. With a flow rate of 1,8 m3/h a...
UOM:  1 * 1 items
Supplier:  Welch by Gardner Denver
Local Article Number:: ILMV411721
Description:   Vacuum Diaphragm Pump, Standard duty diaphragm pump, MP 601 E, No. of stages: 1, Flow rate (l/min): 63 at 50 Hz: 70 at 60 Hz, Flow rate (m³/h): 3,8, Protection class: IP 54, W×D×H (mm): 230×265×169, Weight (kg): 11,2
UOM:  1 * 1 items
Supplier:  Welch by Gardner Denver
Description:   Pumps are constructed with PTFE heads, perfluoroelastomer valves and a PTFE coated molded diaphragm. Includes intake/exhaust hose barbs. DRYFAST® mode...
Catalogue Number: (PALL13158)
Supplier:  Pall Laboratory
Local Article Number:: PALL13158
Description:   This combined vacuum pump and compressed air supply is guaranteed oil and dust-free and helps to keep the entire laboratory spotlessly clean.
UOM:  1 * 1 items
Catalogue Number: (ILMV2546C-02)
Supplier:  Welch by Gardner Denver
Local Article Number:: ILMV2546C-02
Description:   All standard WOB-L piston pumps are recommended for pumping vapors of aqueous solutions including buffers but not for acidic, basic or organic vapors ...
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 0800 734 100.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 10800 734 100.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email Global Science - a Bio-Strategy Company Regulatory Department at Regulatory_Affairs@globalscience.co.nz
-Additional Documentation May be needed to purchase this item. A Global Science representative will contact you if needed.
This product has been blocked by your Organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the Global Science - a Bio-Strategy Company Catalogue Number listed above. If you need further assistance, please call Global Science - a Bio-Strategy Company Customer Service at 0800 734 100.
1 - 6  of 6